accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CCH3

CAS RN 67562-19-0

IUPAC Name: 4,4,5,5-tetramethyl-2-propyl-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-propyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 170.06
Molecular Formula: C9H19BO2
InChi Key: BTSWCGATJFXUHR-UHFFFAOYSA-N
SMILES: CCCB1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results