Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propylboronic acid pinacol ester, 98%, Thermo Scientific™

Catalog No. AAH3044106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3044106 5 g
AAH3044103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3044106 Supplier Thermo Scientific Chemicals Supplier No. H3044106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67562-19-0
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.06
MDL Number MFCD12546187
InChI Key BTSWCGATJFXUHR-UHFFFAOYSA-N
Synonym 1-propylboronic acid pinacol ester, 2-propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-propyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-n-propyl-1,3,2-dioxaborolane
PubChem CID 10103569
IUPAC Name 4,4,5,5-tetramethyl-2-propyl-1,3,2-dioxaborolane
SMILES CCCB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 166°C to 168°C
Flash Point 47°C (117°F)
Quantity 5 g
UN Number UN1993
Formula Weight 170.06
Percent Purity 98%
Chemical Name or Material 1-Propylboronic acid pinacol ester
Hazard Category Warning
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.