accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHHNCH3

CAS RN 67579-81-1

IUPAC Name: N1,N2-dimethylcyclohexane-1,2-diamine
Synonyms: N1,N2-dimethylcyclohexane-1,2-diamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: JRHPOFJADXHYBR-UHFFFAOYNA-N
SMILES: CNC1CCCCC1NC
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)