Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-N,N'-Dimethylcyclohexane-1,2-diamine

Catalog No. 6330895ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-308-95ML 5 mL
63-308-9100ML 100 mL
63-308-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 63-308-95ML Supplier Sigma Aldrich Supplier No. 6330895ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-n,nα-dimethylcyclohexane-1,2-diamine can be used as a ligand in the synthesis of the following products via copper catalyzed c-n coupling reactions:
  • Vinylsulfoximines obtained from nh sulfoximes and vinyl bromides
  • N-arylpyridones obtained via reaction between 2-substituted pyridines and aryl halides
  • N-aryl amines obtained via reaction between amines and aryl iodides/aryl bromides

Specifications

CAS 67579-81-1
Density 0.902 g/mL (at 25°C (literature))
Boiling Point 78°C to 80°C (18 mmHg)
Molecular Formula C8H18N2
Refractive Index n20/D 1.472 (literature)
Linear Formula C8H18N2
MDL Number MFCD03001702
Quantity 5 mL
Molecular Weight (g/mol) 142.24
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.