accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 676-22-2

IUPAC Name: (1E,5E,9E)-cyclododeca-1,5,9-triene
Synonyms: 1,5,9-cyclododecatriene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: ZOLLIQAKMYWTBR-FFWAUJBHSA-N
SMILES: C1C\C=C\CC\C=C\CC\C=C\1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)