Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans,trans-1,5,9-Cyclododecatriene, 97%, stab. with 30-50ppm 4-tert-butylcatechol

Catalog No. AAL0931714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0931714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0931714 Supplier Thermo Scientific Chemicals Supplier No. L0931714

missing translation for 'validMainframeMsg'

CAS: 676-22-2 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-22-2
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00003720
InChI Key ZOLLIQAKMYWTBR-MBKAWSJDSA-N
Synonym 1,5,9-cyclododecatriene, e,e,e, 1,5,9-cyclododecatriene
PubChem CID 20835579
IUPAC Name (5E,9E)-cyclododeca-1,5,9-triene
SMILES C1CC=CCCC=CCCC=C1

Specifications

Melting Point 32°C to 34°C
Density 0.88
Boiling Point 237°C to 238°C
Flash Point 73°C (163°F)
Odor Pungent
Quantity 25 g
UN Number UN1759
Beilstein 969880
Formula Weight 162.28
Percent Purity 97%
Chemical Name or Material trans,trans,trans-1,5,9-Cyclododecatriene, stabilized with 30 to 50ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 211-626-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.