accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 676-58-4

IUPAC Name: chloro(methyl)magnesium
Synonyms: chloromethylmagnesium
Molecular Weight (g/mol): 74.79
Molecular Formula: CH3ClMg
InChi Key: RQNMYNYHBQQZSP-UHFFFAOYSA-M
SMILES: C[Mg]Cl
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results