Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylmagnesium Chloride Solution

Catalog No. 1899018L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
8 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-990-18L 8 L
18-990-1800ML 800 mL
18-990-118LKL 18 L
18-990-125ML 4 x 25 mL
18-990-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 18-990-18L Supplier Sigma Aldrich Supplier No. 1899018L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylmagnesium chloride solution (3 M in THF) undergoes 1,3-addition with dialdose derived nitrones to form N-benzylhydroxylamines. The reductive cleavage of the N-O bond of these N-benzylhydroxylamines results in N-benzylaminosugars, which are key moieties of bioactive compounds. It can also react with Garner type aldehydes to form the corresponding secondary alcohols.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 676-58-4
Density 1.013 g/mL at 25°C
Molecular Formula CH3ClMg
Linear Formula CH3MgCl
MDL Number MFCD00000468
Quantity 8 L
Molecular Weight (g/mol) 74.79 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.