accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PClCl

CAS RN 676-83-5

IUPAC Name: dichloro(methyl)phosphane
Synonyms: dichloro(methyl)phosphane
Molecular Weight (g/mol): 116.91
Molecular Formula: CH3Cl2P
InChi Key: CDPKWOKGVUHZFR-UHFFFAOYSA-N
SMILES: CP(Cl)Cl
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)