CAS RN 67617-35-0
CAS RN 67617-35-0
IUPAC Name:
tert-butyl (2R)-2-amino-4-methylpentanoate
Synonyms:
tert-butyl (2R)-2-amino-4-methylpentanoate
Molecular Weight (g/mol):
187.28
Molecular Formula:
C10H21NO2
InChi Key:
HBEJJYHFTZDAHZ-MRVPVSSYSA-N
SMILES:
CC(C)C[C@@H](N)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Advanced Chem Tech H-D-Leu-OBut.HCl, Advanced ChemTech
CAS: 67617-35-0 Molecular Formula: C10H21NO2 Molecular Weight (g/mol): 187.28 MDL Number: MFCD10568783 InChI Key: HBEJJYHFTZDAHZ-MRVPVSSYSA-N IUPAC Name: tert-butyl (2R)-2-amino-4-methylpentanoate SMILES: CC(C)C[C@@H](N)C(=O)OC(C)(C)C