accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNBr

CAS RN 676170-06-2

IUPAC Name: 3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
Synonyms: 3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
Molecular Weight (g/mol): 278.15
Molecular Formula: C13H12BrNO
InChi Key: TWDVYIPKPMKBCL-UHFFFAOYNA-N
SMILES: BrC1=CC=C2C(NC=C2C2CCC(=O)C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results