CAS RN 676465-94-4
IUPAC Name:
3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol
Synonyms:
3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol
Molecular Weight (g/mol):
250.38
Molecular Formula:
C16H26O2
InChi Key:
DBVBDLSFXMYKIU-UHFFFAOYSA-N
SMILES:
CCC(O)(CC)C1=CC(=CC=C1)C(O)(CC)CC
1
–
1
of
1
results
1
1,3-Bis(3-hydroxy-3-pentyl)benzene 90.0+%, TCI America™
CAS: 676465-94-4 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 InChI Key: DBVBDLSFXMYKIU-UHFFFAOYSA-N PubChem CID: 44630004 IUPAC Name: 3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol SMILES: CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O