accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6780-49-0

IUPAC Name: ethyl N-phenylcarboximidate
Synonyms: ethyl N-phenylcarboximidate
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: DRDBNKYFCOLNQO-UHFFFAOYSA-N
SMILES: CCOC=NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)