accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOH3CNNNOOCH3OOCH3

CAS RN 67817-30-5

IUPAC Name: [3,4,6-tris(acetyloxy)-5-azidooxan-2-yl]methyl acetate
Synonyms: [3,4,6-tris(acetyloxy)-5-azidooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 373.32
Molecular Formula: C14H19N3O9
InChi Key: QKGHBQJLEHAMKJ-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(N=[N+]=[N-])C(OC(C)=O)C1OC(C)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results