Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-alpha-D-galactopyranose, TCI America™
SDP

Catalog No. T1731100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1731100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1731100MG Supplier TCI America Supplier No. T1731100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent for click chemistry applications.

Chemical Identifiers

CAS 67817-30-5
Molecular Formula C14H19N3O9
Molecular Weight (g/mol) 373.32
MDL Number MFCD01076182,MFCD01076182,MFCD01076183,MFCD31814465
InChI Key QKGHBQJLEHAMKJ-UHFFFAOYNA-N
PubChem CID 12821201
IUPAC Name [3,4,6-tris(acetyloxy)-5-azidooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(N=[N+]=[N-])C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 121°C
Color White
Quantity 100 mg
Formula Weight 373.32
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-alpha-D-galactopyranose
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.