CAS RN 6792-31-0
IUPAC Name:
tris(hexafluoroprop-1-ene)
Synonyms:
tris(hexafluoropropylene)
Molecular Weight (g/mol):
450.07
Molecular Formula:
C9F18
InChi Key:
VJRSWIKVCUMTFK-UHFFFAOYSA-N
SMILES:
FC(F)=C(F)C(F)(F)F.FC(F)=C(F)C(F)(F)F.FC(F)=C(F)C(F)(F)F
1
–
1
of
1
results
1
Sigma Aldrich Hexafluoropropene, trimer
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.