accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH3CO

CAS RN 679428-14-9

IUPAC Name: 4-(benzyloxy)-2,6-dimethoxybenzaldehyde
Synonyms: 4-(benzyloxy)-2,6-dimethoxybenzaldehyde
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: YHMXRPWCVSRREV-UHFFFAOYSA-N
SMILES: COC1=CC(OCC2=CC=CC=C2)=CC(OC)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results