Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on Merrifield resin, Thermo Scientific™

Catalog No. AAL1975406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1975406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1975406 Supplier Thermo Scientific Chemicals Supplier No. L1975406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 679428-14-9
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD02683435
InChI Key YHMXRPWCVSRREV-UHFFFAOYSA-N
Synonym 4-benzyloxy-2,6-dimethoxybenzaldehyde, 4-benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on merrifield resin, fdmp resin, syringaldehyde, polymer-bound, stratospheres pl-fdmp resin, 2,6-dimethoxy-4-phenylmethoxy-benzaldehyde, 4-formyl-3,5-dimethoxyphenoxymethyl polystyrene, benzaldehyde, 2,6-dimethoxy-4-phenylmethoxy, 4-hydroxy-2,6-dimethoxybenzaldehyde, polymer-bound
PubChem CID 4607337
IUPAC Name 2,6-dimethoxy-4-phenylmethoxybenzaldehyde
SMILES COC1=CC(OCC2=CC=CC=C2)=CC(OC)=C1C=O

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Chemical Name or Material 4-Benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0 to 1.5mmol/g on Merrifield resin
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.