accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOH2NOHPOHOHO

CAS RN 67952-32-3

IUPAC Name: 1-aminopropan-2-ol; phosphoric acid
Synonyms: 1-amino-2-propanol; phosphoric acid
Molecular Weight (g/mol): 173.11
Molecular Formula: C3H12NO5P
InChi Key: NSTLSXSJOBWQLC-UHFFFAOYNA-N
SMILES: CC(O)CN.OP(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results