Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanolamine Phosphate 98.0+%, TCI America™
SDP

Catalog No. I022125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I022125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I022125G Supplier TCI America Supplier No. I022125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67952-32-3
Molecular Formula C3H12NO5P
Molecular Weight (g/mol) 173.11
MDL Number MFCD00070562
InChI Key NSTLSXSJOBWQLC-UHFFFAOYNA-N
IUPAC Name 1-aminopropan-2-ol; phosphoric acid
SMILES CC(O)CN.OP(O)(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 248.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropanolamine Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.