accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67953-08-6

ClH2NOH2NNOOOHO

CAS RN 67953-08-6

IUPAC Name: N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
Synonyms: N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
Molecular Weight (g/mol): 303.7
Molecular Formula: C11H14ClN3O5
InChi Key: WMRCHNNHOJNRSL-UHFFFAOYNA-N
SMILES: [Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results