accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 679806-84-9

IUPAC Name: 4-(pyrimidin-2-yl)benzoyl chloride
Synonyms: 4-(pyrimidin-2-yl)benzoyl chloride
Molecular Weight (g/mol): 218.64
Molecular Formula: C11H7ClN2O
InChi Key: AQFRTZZTRGGBKO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results