Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyrimidin-2-ylbenzoyl chloride, TECH, Thermo Scientific™

Catalog No. CC56202DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56202DA 1 g Amber Glass Bottle
CC56202CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC56202DA Supplier Thermo Scientific Chemicals Supplier No. CC56202DA
Add to Cart

Chemical Identifiers

CAS 679806-84-9
Molecular Formula C11H7ClN2O
Molecular Weight (g/mol) 218.64
MDL Number MFCD09064974
InChI Key AQFRTZZTRGGBKO-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride
PubChem CID 21092945
IUPAC Name 4-pyrimidin-2-ylbenzoyl chloride
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 218.64
Percent Purity 90%
Chemical Name or Material 4-Pyrimidin-2-ylbenzoyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.