accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCH3CH3H3CCH3(E)(E)(E)(E)

CAS RN 68-26-8

IUPAC Name: (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol
Synonyms: vitamin A anatola A α-vi-pel alcovit A vio-A retrovitamin A thalasphere dofsol veroftal ro-a-vit
Molecular Weight (g/mol): 286.46
Molecular Formula: C20H30O
InChi Key: FPIPGXGPPPQFEQ-OVSJKPMPSA-N
SMILES: C\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Molecular Weight (g/mol) 
  • (7)
  • (3)
Physical Form 
  • (4)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (7)
  • (3)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)