Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinol, MP Biomedicals™

Catalog No. ICN19026890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19026890 500 mg
ICN19026825 25 mg
ICN19026880 100 mg
ICN19026883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19026890 Supplier MP Biomedicals, Inc Supplier No. 0219026890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68-26-8
Molecular Formula C20H30O
Molecular Weight (g/mol) 286.459
MDL Number MFCD00001552
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
PubChem CID 445354
ChEBI CHEBI:17336
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C

Specifications

Color Orange
Quantity 500 mg
Formula Weight 286.5
Physical Form Powder
Chemical Name or Material Retinol, Molecular biology
EINECSNumber 200-683-7
Recommended Storage Store at 0°C under nitrogen, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.