accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl

CAS RN 68-36-0

IUPAC Name: 1,4-bis(trichloromethyl)benzene
Synonyms: P-DI(trichloromethyl)benzene
Molecular Weight (g/mol): 312.82
Molecular Formula: C8H4Cl6
InChi Key: OTEKOJQFKOIXMU-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C1=CC=C(C=C1)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results