Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha,alpha',alpha',alpha'-Hexachloro-p-xylene 98.0+%, TCI America™
SDP

Catalog No. H006425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H006425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H006425G Supplier TCI America Supplier No. H006425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68-36-0
Molecular Formula C8H4Cl6
Molecular Weight (g/mol) 312.82
MDL Number MFCD00000791
InChI Key OTEKOJQFKOIXMU-UHFFFAOYSA-N
Synonym 1,4-bis trichloromethyl benzene, bitriben, chloksil, chloxil, chloxyl, hexichol, khloksil, khloxil, cloxil, hetol
PubChem CID 6233
IUPAC Name 1,4-bis(trichloromethyl)benzene
SMILES ClC(Cl)(Cl)C1=CC=C(C=C1)C(Cl)(Cl)Cl

Specifications

Melting Point 108°C
Color White-Yellow
Boiling Point 312°C
Quantity 25 g
UN Number 1759
Formula Weight 312.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha,alpha′,alpha′,alpha′-Hexachloro-p-xylene
EINECSNumber (3)-0049
RTECSNumber ZE4655000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.