accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNHI

CAS RN 68007-08-9

IUPAC Name: 1H-imidazole hydroiodide
Synonyms: imidazole hydroiodide
Molecular Weight (g/mol): 195.99
Molecular Formula: C3H5IN2
InChi Key: JBOIAZWJIACNJF-UHFFFAOYSA-N
SMILES: I.N1C=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results