Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole Hydroiodide (Low water content) 98.0+%, TCI America™
SDP

Catalog No. I09705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09705G 5 g
I09701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09705G Supplier TCI America Supplier No. I09705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68007-08-9
Molecular Formula C3H5IN2
Molecular Weight (g/mol) 195.991
InChI Key JBOIAZWJIACNJF-UHFFFAOYSA-N
Synonym Imidazolium Iodide
PubChem CID 12203051
IUPAC Name 1H-imidazole;hydroiodide
SMILES C1=CN=CN1.I

Specifications

Quantity 5 g
Formula Weight 195.99
Percent Purity ≥98.0% (T)
Chemical Name or Material Imidazole Hydroiodide (Low water content)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.