accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 680569-18-0

H3CNBrOOH3C

CAS RN 680569-18-0

IUPAC Name: methyl 6-bromo-1-methyl-1H-indole-2-carboxylate
Synonyms: methyl 6-bromo-1-methylindole-2-carboxylate
Molecular Weight (g/mol): 268.11
Molecular Formula: C11H10BrNO2
InChi Key: RCDWEKOOOHATJC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=C(Br)C=C2N1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results