Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-bromo-1-methylindole-2-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH6365506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6365506 5 g
AAH6365503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6365506 Supplier Thermo Scientific Chemicals Supplier No. H6365506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680569-18-0
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD07364734
InChI Key RCDWEKOOOHATJC-UHFFFAOYSA-N
Synonym methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
PubChem CID 4777911
IUPAC Name methyl 6-bromo-1-methylindole-2-carboxylate
SMILES CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 268.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 6-bromo-1-methylindole-2-carboxylate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.