accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 6807-11-0

IUPAC Name: 2-(4-methylphenoxy)ethyl acetate
Synonyms: 2-P-cresyloxyethyl acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: ZJMLZGKLUMJGSL-UHFFFAOYSA-N
SMILES: CC(=O)OCCOC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results