Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Acetoxyethoxy)toluene 98.0+%, TCI America™
SDP

Catalog No. A081910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A081910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A081910G Supplier TCI America Supplier No. A081910G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6807-11-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00059337
InChI Key ZJMLZGKLUMJGSL-UHFFFAOYSA-N
Synonym Acetic Acid 2-p-Tolyloxyethyl Ester, Ethylene Glycol Mono-p-tolyl Ether Acetate, 2-p-Tolyloxyethyl Acetate
PubChem CID 81258
IUPAC Name 2-(4-methylphenoxy)ethyl acetate
SMILES CC(=O)OCCOC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 10 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(2-Acetoxyethoxy)toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.