accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OHOH

CAS RN 6807-17-6

IUPAC Name: 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol
Synonyms: 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol
Molecular Weight (g/mol): 270.37
Molecular Formula: C18H22O2
InChi Key: VHLLJTHDWPAQEM-UHFFFAOYSA-N
SMILES: CC(C)CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results