Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-(1,3-Dimethylbutylidene)diphenol 98.0+%, TCI America™
SDP

Catalog No. D326725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326725G 25 g
D3267500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D326725G Supplier TCI America Supplier No. D326725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6807-17-6
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.372
InChI Key VHLLJTHDWPAQEM-UHFFFAOYSA-N
Synonym 2,2-Bis(4-hydroxyphenyl)-4-methylpentane
PubChem CID 81259
IUPAC Name 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol
SMILES CC(C)CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 270.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-(1,3-Dimethylbutylidene)diphenol
EINECSNumber (4)-1709
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.