accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 681-99-2

IUPAC Name: tributyl(isocyanato)stannane
Synonyms: tributyl(isocyanato)stannane
Molecular Weight (g/mol): 334.09
Molecular Formula: C13H29NOSn
InChi Key: KYQISWRYYMJYEP-UHFFFAOYSA-N
SMILES: N=C=O.CCCC[SnH](CCCC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)