accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHBrHCl

CAS RN 68104-62-1

IUPAC Name: 1-(4-bromophenyl)piperazine hydrochloride
Synonyms: 1-(4-bromophenyl)piperazine hydrochloride
Molecular Weight (g/mol): 277.59
Molecular Formula: C10H14BrClN2
InChi Key: YDVSFRZKQMQPJD-UHFFFAOYSA-N
SMILES: Cl.BrC1=CC=C(C=C1)N1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results