Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)piperazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B36305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36305G 5 g
B36301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36305G Supplier TCI America Supplier No. B36305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68104-62-1
Molecular Formula C10H14BrClN2
Molecular Weight (g/mol) 277.59
MDL Number MFCD00674134
InChI Key YDVSFRZKQMQPJD-UHFFFAOYSA-N
PubChem CID 14422838
IUPAC Name 1-(4-bromophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)Br.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 277.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)piperazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.