accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 6812-14-2

IUPAC Name: 1H,3H-anthra[2,3-c]furan-1,3-dione
Synonyms: anthra[2,3-c]furan-1,3-dione
Molecular Weight (g/mol): 248.24
Molecular Formula: C16H8O3
InChi Key: AJXNLGUENUIIRW-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC3=CC4=CC=CC=C4C=C3C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results