Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Anthracenedicarboxylic Anhydride 93.0+%, TCI America™
SDP

Catalog No. A1834500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1834500MG 500 mg
A1834100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1834500MG Supplier TCI America Supplier No. A1834500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6812-14-2
Molecular Formula C16H8O3
Molecular Weight (g/mol) 248.237
InChI Key AJXNLGUENUIIRW-UHFFFAOYSA-N
PubChem CID 11096934
IUPAC Name naphtho[2,3-f][2]benzofuran-1,3-dione
SMILES C1=CC=C2C=C3C=C4C(=CC3=CC2=C1)C(=O)OC4=O

Specifications

Color Yellow
Quantity 500 mg
Formula Weight 248.24
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Anthracenedicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.