accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68133-98-2

IUPAC Name: hydrogen [(4-nitrophenyl)methyl](propyl)amine chloride
Synonyms: hydrogen [(4-nitrophenyl)methyl](propyl)amine chloride
Molecular Weight (g/mol): 230.69
Molecular Formula: C10H15ClN2O2
InChi Key: LWISLZIFEARHJI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCNCC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results