accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNHOOHOOH(E/Z)

CAS RN 681806-46-2

IUPAC Name: 2-[(4,5-dihydro-1H-imidazol-2-yl)methyl]-1-methyl-2,3-dihydro-1H-isoindole; but-2-enedioic acid
Synonyms: 2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole; butenedioic acid
Molecular Weight (g/mol): 331.37
Molecular Formula: C17H21N3O4
InChi Key: DDIQGSUEJOOQQQ-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CC1N(CC2=NCCN2)CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)