accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSSNONOO

CAS RN 68181-17-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulfanyl)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulfanyl)propanoate
Molecular Weight (g/mol): 312.36
Molecular Formula: C12H12N2O4S2
InChi Key: JWDFQMWEFLOOED-UHFFFAOYSA-N
SMILES: O=C(CCSSC1=CC=CC=N1)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)