Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich D-(-)-Alpha-Phenylglycine

Catalog No. P2548525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2548525G 5 mg
P25485100G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2548525G Supplier Sigma Aldrich Supplier No. P2548525G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

D-(-)-Alpha-Phenylglycine

Specifications

CAS 68181-17-9
Melting Point 84°C to 86°C
Molecular Formula C12H12N2O4S2
Linear Formula C12H12N2O4S2
MDL Number MFCD00009636
Quantity 5 mg
Synonym (R)-(-)-2-Phenylglycine; D-2-Phenylglycine; R-(-)-alpha-Aminophenylacetic acid
Molecular Weight (g/mol) 312.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.