accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFFFCH3NNH3C

CAS RN 681856-28-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; trifluoro(trifluoromethyl)boranuide
Synonyms: 1-ethyl-3-methylimidazolium; trifluoro(trifluoromethyl)boranuide
Molecular Weight (g/mol): 247.98
Molecular Formula: C7H11BF6N2
InChi Key: YTANZWHOSGQGAP-UHFFFAOYSA-N
SMILES: CCN1C=C[N+](C)=C1.F[B-](F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results