Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium Trifluoro(trifluoromethyl)borate 98.0+%, TCI America™
SDP

Catalog No. E08365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08365G Supplier TCI America Supplier No. E08365G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 681856-28-0
Molecular Formula C7H11BF6N2
Molecular Weight (g/mol) 247.979
InChI Key YTANZWHOSGQGAP-UHFFFAOYSA-N
PubChem CID 44630031
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;trifluoro(trifluoromethyl)boranuide
SMILES [B-](C(F)(F)F)(F)(F)F.CCN1C=C[N+](=C1)C

Specifications

Melting Point -20°C
Color Yellow
Quantity 5 g
Formula Weight 247.98
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Trifluoro(trifluoromethyl)borate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.