accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NN(E)

CAS RN 68189-23-1

IUPAC Name: 4-[(E)-(2,2-diphenylhydrazin-1-ylidene)methyl]-N,N-diethylaniline
Synonyms: 4-[(E)-(2,2-diphenylhydrazin-1-ylidene)methyl]-N,N-diethylaniline
Molecular Weight (g/mol): 343.47
Molecular Formula: C23H25N3
InChi Key: YGBCLRRWZQSURU-LYBHJNIJSA-N
SMILES: CCN(CC)C1=CC=C(\C=N\N(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results