Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethylamino)benzaldehyde Diphenylhydrazone 98.0+%, TCI America™
SDP

Catalog No. D29521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29521G Supplier TCI America Supplier No. D29521G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68189-23-1
Molecular Formula C23H25N3
Molecular Weight (g/mol) 343.47
MDL Number MFCD00143309
InChI Key YGBCLRRWZQSURU-LYBHJNIJSA-N
Synonym 4-diethylamino benzaldehyde diphenylhydrazone, 4-diethylamino benzaldehyde-1,1-diphenylhydrazone, 4-2,2-diphenylhydrazono methyl-n,n-diethylaniline, 4-e-diphenylhydrazinylidene methyl-n,n-diethylaniline, 4-diethylamino benzaldehyde-n,n-diphenylhydrazone, 4-diethylaminobenzaldehyde diphenylhydrazone, p-diethylami-nobenzaldehyde diphenylhydrazone, 4-diethylamino benzaldehyde diphenylhydrazone #, 4-diethylaminobenzaldehyde n,n'-diphenylhydrazone, 4-diethylaminobenzaldehyde n,n-diphenylhydrazone
PubChem CID 9580366
IUPAC Name 4-[(E)-(2,2-diphenylhydrazin-1-ylidene)methyl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(\C=N\N(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 94°C
Color Yellow
Quantity 1 g
Formula Weight 343.47
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Diethylamino)benzaldehyde Diphenylhydrazone
RTECSNumber CU5612900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.