accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiOCH3OH3COH3C

CAS RN 682-01-9

IUPAC Name: tetrapropyl silicate
Synonyms: tetrapropoxy silane
Molecular Weight (g/mol): 264.44
Molecular Formula: C12H28O4Si
InChi Key: ZQZCOBSUOFHDEE-UHFFFAOYSA-N
SMILES: CCCO[Si](OCCC)(OCCC)OCCC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)