Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrapropoxysilane, 99+%, Thermo Scientific™

Catalog No. AC250430250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC250430250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250430250 Supplier Thermo Scientific Chemicals Supplier No. 250430250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 682-01-9
Molecular Formula C12H28O4Si
Molecular Weight (g/mol) 264.437
InChI Key ZQZCOBSUOFHDEE-UHFFFAOYSA-N
Synonym tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane
PubChem CID 69627
IUPAC Name tetrapropyl silicate
SMILES CCCO[Si](OCCC)(OCCC)OCCC

Specifications

Color Colorless
Density 0.916g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Refractive Index 1.3995 to 1.4015 (20°C, 589nm)
Linear Formula Si(OCH2CH2CH3)4
Quantity 25 mL
Beilstein 01, 355
Formula Weight 264.44
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material Tetrapropoxysilane, 99+%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.